Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spink8
Entrez Gene ID
301016
Gene Name
serine peptidase inhibitor, Kazal type 8
Gene Aliases
RGD1307150
Chromosome Location
Chr.8: 117906093 - 117917821 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106860.1 NP_001100330.1 1 - 2 124 63

View Full Product Details