Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gcm1
Entrez Gene ID
29394
Gene Name
glial cells missing homolog 1 (Drosophila)
Gene Aliases
Gcma
Chromosome Location
Chr.8: 85355766 - 85369002 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017186.2 NP_058882.1 1 - 2 43 75
GenBank mRNA AF081557.1 - 1 - 2 44 75

View Full Product Details