Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sec22c
Entrez Gene ID
687022
Gene Name
SEC22 vesicle trafficking protein homolog C (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.8: 130319682 - 130339245 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766703.1 XP_008764925.1 5 - 6 1643 87

View Full Product Details