Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa5
Entrez Gene ID
301013
Gene Name
shisa family member 5
Gene Aliases
-
Chromosome Location
Chr.8: 117781014 - 117795923 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243762.2 XP_006243824.1 1 - 2 409 100

View Full Product Details