The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105711.1 | NP_001099181.1 | 5 - 6 | 2746 | 91 |
GenBank mRNA | M92075.1 | - | 6 - 7 | 2739 | 91 |