The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012001.1 | NP_001012001.1 | 7 - 8 | 994 | 117 |
XM_006243926.2 | XP_006243988.1 | 7 - 8 | 1201 | 117 | |
GenBank mRNA | BC085884.1 | - | 7 - 8 | 994 | 117 |