Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc7
Entrez Gene ID
316098
Gene Name
exosome component 7
Gene Aliases
-
Chromosome Location
Chr.8: 132204628 - 132229968 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100725.1 NP_001094195.1 2 - 3 161 91
GenBank mRNA BF561514.1 - 2 - 3 167 91
FQ226090.1 - 2 - 3 166 91

View Full Product Details