Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Slc25a36
- Entrez Gene ID
-
501039
- Gene Name
- solute carrier family 25 (pyrimidine nucleotide carrier ), member 36
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 104962569 - 104995932 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_576451.6 |
XP_576451.4 |
2 - 3 |
414 |
76 |
- Gene Symbol
- Slc25a36l1
- Entrez Gene ID
-
364991
- Gene Name
- solute carrier family 25 (pyrimidine nucleotide carrier ), member 36-like 1
- Gene Aliases
-
RGD1561851
- Chromosome Location
-
Chr.19: 37020220 - 37021997 on Build Rnor_6.0
View Full Product Details