Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gnat1
Entrez Gene ID
363143
Gene Name
guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.8: 116433302 - 116438038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108780.2 NP_001102250.2 4 - 5 511 66

View Full Product Details