The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048243.2 | NP_001041708.2 | 4 - 5 | 294 | 121 |
XM_006242661.2 | XP_006242723.1 | 4 - 5 | 475 | 121 | |
GenBank mRNA | BC157818.1 | - | 3 - 4 | 197 | 121 |
CB713235.1 | - | 4 - 5 | 294 | 121 |