Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Folr4
Entrez Gene ID
690037
Gene Name
folate receptor 4, delta
Gene Aliases
-
Chromosome Location
Chr.8: 13416418 - 13420117 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001072998.4 XP_001072998.2 1 - 2 164 57

View Full Product Details