Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfm2
Entrez Gene ID
313783
Gene Name
olfactomedin 2
Gene Aliases
-
Chromosome Location
Chr.8: 21684302 - 21761984 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015017.1 NP_001015017.1 2 - 3 446 71
XM_006242641.2 XP_006242703.1 3 - 4 738 71
XM_006242642.2 XP_006242704.1 2 - 3 2015 71
XM_006242643.1 XP_006242705.1 2 - 3 258 71
GenBank mRNA BC092647.1 - 2 - 3 446 71

View Full Product Details