The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172324.1 | NP_758827.1 | 5 - 6 | 754 | 59 |
XM_006242616.2 | XP_006242678.1 | 5 - 6 | 1098 | 59 | |
XM_008765920.1 | XP_008764142.1 | 5 - 6 | 941 | 59 | |
GenBank mRNA | AB097198.1 | - | 5 - 6 | 754 | 59 |