Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rdh8
Entrez Gene ID
690953
Gene Name
retinol dehydrogenase 8 (all-trans)
Gene Aliases
-
Chromosome Location
Chr.8: 21835103 - 21842374 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168593.1 NP_001162065.1 5 - 6 722 83

View Full Product Details