The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008317.1 | NP_001008318.1 | 11 - 12 | 1198 | 63 |
GenBank mRNA | AY158662.1 | - | 11 - 12 | 1198 | 63 |
FQ211910.1 | - | - | 1292 | 63 | |
FQ216126.1 | - | 11 - 12 | 1253 | 63 | |
FQ227188.1 | - | 11 - 12 | 1259 | 63 | |
FQ234126.1 | - | 11 - 12 | 1258 | 63 | |
FQ234429.1 | - | 11 - 12 | 1298 | 63 |