Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erp44
Entrez Gene ID
298066
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
BWK4, Txndc4
Chromosome Location
Chr.5: 63937866 - 64030827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008317.1 NP_001008318.1 1 - 2 137 151
GenBank mRNA AY158662.1 - 1 - 2 137 151
FQ211910.1 - - 232 150
FQ216126.1 - 1 - 2 191 151
FQ227188.1 - 1 - 2 197 151
FQ234126.1 - 1 - 2 198 151
FQ234429.1 - 1 - 2 238 151

View Full Product Details