The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107931.2 | NP_001101401.1 | 1 - 2 | 407 | 116 |
GenBank mRNA | EU487255.1 | - | 1 - 2 | 407 | 116 |