Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctsr
Entrez Gene ID
290975
Gene Name
cathepsin R
Gene Aliases
Cts6
Chromosome Location
Chr.17: 3885684 - 3894447 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175581.3 NP_783171.2 2 - 3 201 113
GenBank mRNA BC097484.1 - 2 - 3 201 113

View Full Product Details