Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab24
Entrez Gene ID
361208
Gene Name
RAB24, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.17: 9837259 - 9839464 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015023.1 NP_001015023.1 2 - 3 612 55
XM_006253632.2 XP_006253694.1 2 - 3 503 55
GenBank mRNA BC091184.1 - 2 - 3 612 55

View Full Product Details