Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Selm
Entrez Gene ID
498398
Gene Name
selenoprotein M
Gene Aliases
RGD1565037
Chromosome Location
Chr.14: 83752393 - 83754975 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115013.1 NP_001108485.1 4 - 5 330 61
GenBank mRNA BI282694.1 - 1 - 2 61
CB607044.1 - 4 - 5 302 61
FQ223498.1 - 4 - 5 285 61

View Full Product Details