Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpr21
Entrez Gene ID
689917
Gene Name
G protein-coupled receptor 21
Gene Aliases
-
Chromosome Location
Chr.3: 21764377 - 21767300 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109554.1 NP_001103024.1 1 - 2 114 62

View Full Product Details