Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Olfml2a
Entrez Gene ID
296708
Gene Name
olfactomedin-like 2A
Gene Aliases
-
Chromosome Location
Chr.3: 23223468 - 23259318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106572.1 NP_001100042.1 4 - 5 847 113
XM_006234074.2 XP_006234136.1 5 - 6 1055 113

View Full Product Details