The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106572.1 | NP_001100042.1 | 6 - 7 | 1287 | 126 |
XM_006234074.2 | XP_006234136.1 | 7 - 8 | 1495 | 126 |