The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134580.1 | NP_001128052.1 | 9 - 10 | 864 | 63 |
XM_008761762.1 | XP_008759984.1 | 6 - 7 | 2710 | 63 | |
XM_008761763.1 | XP_008759985.1 | 7 - 8 | 3360 | 63 | |
XM_008761764.1 | XP_008759986.1 | 6 - 7 | 3176 | 63 | |
XM_008761765.1 | XP_008759987.1 | 8 - 9 | 3480 | 63 | |
XM_008761766.1 | XP_008759988.1 | 5 - 6 | 2650 | 63 | |
XM_008761767.1 | XP_008759989.1 | 5 - 6 | 3116 | 63 | |
XM_008761768.1 | XP_008759990.1 | 5 - 6 | 2160 | 63 | |
XM_008761769.1 | XP_008759991.1 | 6 - 7 | 2616 | 63 |