Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March7
Entrez Gene ID
311059
Gene Name
membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase
Gene Aliases
Axot
Chromosome Location
Chr.3: 46284962 - 46325230 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012087.1 NP_001012087.1 3 - 4 533 67
XM_008761841.1 XP_008760063.1 3 - 4 1224 67
XM_008761842.1 XP_008760064.1 2 - 3 356 67
XM_008761843.1 XP_008760065.1 2 - 3 356 67
XM_008761844.1 XP_008760066.1 3 - 4 814 67
GenBank mRNA BC083842.1 - 3 - 4 533 67

View Full Product Details