Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Galnt5
Entrez Gene ID
83627
Gene Name
polypeptide N-acetylgalactosaminyltransferase 5
Gene Aliases
-
Chromosome Location
Chr.3: 44025964 - 44072073 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031796.1 NP_113984.1 4 - 5 1961 132
GenBank mRNA AF049344.1 - 4 - 5 1961 132

View Full Product Details