Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cacnb4
Entrez Gene ID
58942
Gene Name
calcium channel, voltage-dependent, beta 4 subunit
Gene Aliases
-
Chromosome Location
Chr.3: 37950846 - 38092383 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105733.1 NP_001099203.1 1 - 2 304 152

View Full Product Details