Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Coq2
Entrez Gene ID
498332
Gene Name
coenzyme Q2 4-hydroxybenzoate polyprenyltransferase
Gene Aliases
RGD1306722
Chromosome Location
Chr.14: 10581104 - 10601093 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044255.1 NP_001037720.1 4 - 5 635 73
XM_006250675.2 XP_006250737.1 4 - 5 667 73
GenBank mRNA BC099827.1 - 4 - 5 635 73

View Full Product Details