Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Enoph1
Entrez Gene ID
305177
Gene Name
enolase-phosphatase 1
Gene Aliases
RGD1309016
Chromosome Location
Chr.14: 11172359 - 11198273 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009391.1 NP_001009391.1 5 - 6 737 78
XM_008770023.1 XP_008768245.1 5 - 6 787 78
GenBank mRNA BC087697.1 - 5 - 6 737 78

View Full Product Details