Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmp1
Entrez Gene ID
25312
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
DENTMAT
Chromosome Location
Chr.14: 6889851 - 6923986 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203493.3 NP_987089.2 1 - 2 66 92
XM_006250622.2 XP_006250684.1 5 - 6 2639 92
XM_008769994.1 XP_008768216.1 1 - 2 337 92
XM_008769995.1 XP_008768217.1 4 - 5 756 92
GenBank mRNA BC129082.1 - 1 - 2 66 92
L11354.1 - 2 - 3 200 92

View Full Product Details