Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Paqr3
Entrez Gene ID
305203
Gene Name
progestin and adipoQ receptor family member III
Gene Aliases
-
Chromosome Location
Chr.14: 14107518 - 14126464 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012033.1 NP_001012033.1 2 - 3 464 57
XM_008770025.1 XP_008768247.1 2 - 3 526 57
GenBank mRNA BC079418.1 - 2 - 3 464 57

View Full Product Details