Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agpat9
Entrez Gene ID
305166
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 9
Gene Aliases
-
Chromosome Location
Chr.14: 10343290 - 10396561 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025670.1 NP_001020841.1 4 - 5 852 93
XM_008770019.1 XP_008768241.1 5 - 6 849 93
XM_008770020.1 XP_008768242.1 5 - 6 1278 93
XM_008770021.1 XP_008768243.1 5 - 6 977 93
GenBank mRNA BC097362.1 - 4 - 5 852 93

View Full Product Details