Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wapal
Entrez Gene ID
290577
Gene Name
wings apart-like homolog (Drosophila)
Gene Aliases
RGD1308960
Chromosome Location
Chr.16: 11029449 - 11108867 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252723.2 XP_006252785.1 10 - 11 2809 103
XM_006252724.2 XP_006252786.1 10 - 11 2921 103
XM_006252725.2 XP_006252787.1 10 - 11 2797 103
XM_006252726.2 XP_006252788.1 9 - 10 2656 103

View Full Product Details