The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030849.1 | NP_110476.1 | 11 - 12 | 1701 | 65 |
GenBank mRNA | D17667.1 | - | 10 - 11 | 1476 | 65 |
D38082.1 | - | 10 - 11 | 1558 | 65 | |
S75359.1 | - | 11 - 12 | 1701 | 65 |