The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109118.1 | NP_001102588.1 | 6 - 7 | 759 | 75 |
XM_006253779.2 | XP_006253841.1 | 6 - 7 | 705 | 75 |