Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb3
Entrez Gene ID
499313
Gene Name
zinc finger and BTB domain containing 3
Gene Aliases
RGD1561140
Chromosome Location
Chr.1: 224998100 - 225000489 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109162.2 NP_001102632.1 1 - 2 150 64
GenBank mRNA BC160822.1 - 1 - 2 150 64

View Full Product Details