The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025415.1 | NP_001020586.1 | 1 - 1 | 1151 | 99 |
XM_006231291.2 | XP_006231353.1 | 2 - 2 | 3752 | 99 | |
GenBank mRNA | BC097964.1 | - | 1 - 1 | 1151 | 99 |