Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf6l
Entrez Gene ID
309194
Gene Name
TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor
Gene Aliases
-
Chromosome Location
Chr.1: 224974238 - 224986828 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107575.1 NP_001101045.1 3 - 4 312 88
XM_006230979.2 XP_006231041.1 3 - 4 531 88
XM_006230981.2 XP_006231043.1 3 - 4 474 88
XM_006230982.2 XP_006231044.1 4 - 5 401 88
XM_006230983.2 XP_006231045.1 3 - 4 312 88
XM_008760224.1 XP_008758446.1 3 - 4 783 88
GenBank mRNA BC168911.1 - 3 - 4 427 88

View Full Product Details