The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108513.1 | NP_001101983.1 | 1 - 2 | 165 | 111 |
XM_006230833.1 | XP_006230895.1 | 1 - 2 | 165 | 111 | |
GenBank mRNA | BC088464.1 | - | 1 - 2 | 129 | 111 |