Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
F9
Entrez Gene ID
24946
Gene Name
coagulation factor IX
Gene Aliases
-
Chromosome Location
Chr.X: 143097560 - 143141786 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031540.1 NP_113728.1 4 - 5 370 76
GenBank mRNA FM036569.1 - 4 - 5 334 76

View Full Product Details