The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029927.1 | NP_001025098.1 | 12 - 13 | 1346 | 92 |
XM_006236511.2 | XP_006236573.2 | 12 - 13 | 1822 | 92 | |
XM_008762915.1 | XP_008761137.1 | 14 - 15 | 2131 | 92 | |
GenBank mRNA | BC097471.1 | - | 12 - 13 | 1346 | 92 |