Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slitrk3
Entrez Gene ID
310519
Gene Name
SLIT and NTRK-like family, member 3
Gene Aliases
-
Chromosome Location
Chr.2: 170449182 - 170461513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107683.1 NP_001101153.1 1 - 2 314 74

View Full Product Details