Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Msto1
Entrez Gene ID
295237
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
RGD1306110
Chromosome Location
Chr.2: 188212211 - 188216482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106443.1 NP_001099913.1 2 - 3 269 67

View Full Product Details