Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agk
Entrez Gene ID
502749
Gene Name
acylglycerol kinase
Gene Aliases
RGD1562046
Chromosome Location
Chr.4: 68483336 - 68561518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127497.1 NP_001120969.1 2 - 3 160 76
XM_006236363.2 XP_006236425.1 2 - 3 462 76
XM_008762821.1 XP_008761043.1 2 - 3 557 76

View Full Product Details