Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Theg
Entrez Gene ID
299599
Gene Name
theg spermatid protein
Gene Aliases
-
Chromosome Location
Chr.7: 13013521 - 13023666 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013102.1 NP_001013120.1 1 - 2 336 61
XM_006240857.2 XP_006240919.1 1 - 2 356 61
XM_006240858.2 XP_006240920.1 1 - 2 356 61
GenBank mRNA BC083921.1 - 1 - 2 336 61

View Full Product Details