Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Theg
Entrez Gene ID
299599
Gene Name
theg spermatid protein
Gene Aliases
-
Chromosome Location
Chr.7: 13013521 - 13023666 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240857.2 XP_006240919.1 7 - 8 954 119
XM_006240858.2 XP_006240920.1 6 - 7 882 119

View Full Product Details