Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gipc3
Entrez Gene ID
500789
Gene Name
GIPC PDZ domain containing family, member 3
Gene Aliases
RGD1563255
Chromosome Location
Chr.7: 11245875 - 11250708 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109282.1 NP_001102752.1 1 - 2 186 124

View Full Product Details