Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dapk3
Entrez Gene ID
64391
Gene Name
death-associated protein kinase 3
Gene Aliases
Dapkl
Chromosome Location
Chr.7: 11392436 - 11400805 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC062076.1 - 1 - 2 80 115

View Full Product Details