Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mex3d
Entrez Gene ID
299613
Gene Name
mex-3 RNA binding family member D
Gene Aliases
Rkhd1
Chromosome Location
Chr.7: 12191902 - 12198841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241041.2 XP_006241103.2 1 - 2 730 86

View Full Product Details