The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133308.2 | NP_579842.2 | 3 - 4 | 2127 | 72 |
GenBank mRNA | AF440691.2 | - | 3 - 4 | 2127 | 72 |